Learn More
Alkali Scientific Stainless Steel Canister Frames for CHARTERMED Blood Bag # 7005 2, 7303 2 & 7039 2 (2 Canisters)
SDP
Catalog No. 5000662508
Encompass_Preferred
Canister Frames are constructed of corrosion restant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Frame for 2 CHARTERMED C 7005 Aluminun Canisters Blood Bag Number: 7005 2 7303 2 7039 2 Dimensions (D x H x L): 8 5/8 x 15 1/2 x 9/16″ 212 x 393 x 15mm Canister: C 7005 Platform Dividers P20 7005 P24 7005 P31 7005 P39 7005
Catalog No. 50-006-62508 Supplier Alkali Scientific Supplier No. FC70052
Canister Frames are constructed of corrosion restant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Frame for 2 CHARTERMED C 7005 Aluminun Canisters Blood Bag Number: 7005 2 7303 2 7039 2 Dimensions (D x H x L): 8 5/8 x 15 1/2 x 9/16″ 212 x 393 x 15mm Canister: C 7005 Platform Dividers P20 7005 P24 7005 P31 7005 P39 7005