Learn More
Alkali Scientific Stainless Steel Canister Frames for CHARTERMED Blood Bag # 7120 2 (5 Canisters)
SDP
Catalog No. 5000662982
Encompass_Preferred
Canister Frames are constructed of corrosion restant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Frame for 5 CHARTERMED C 7120 Aluminum Canisters Blood Bag Number: 7120 2 Dimensions (D x H x L): 4 5/16 x 22 1/16 x 9/16″ 110 x 561 x 15mm Canister: C 7120 Platform Dividers P20 7120 P24 7120 P31 7120 P39 7120
Catalog No. 50-006-62982 Supplier Alkali Scientific Supplier No. FC71205
Canister Frames are constructed of corrosion restant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Frame for 5 CHARTERMED C 7120 Aluminum Canisters Blood Bag Number: 7120 2 Dimensions (D x H x L): 4 5/16 x 22 1/16 x 9/16″ 110 x 561 x 15mm Canister: C 7120 Platform Dividers P20 7120 P24 7120 P31 7120 P39 7120