Learn More
Alkali Scientific Stainless Steel Canister Frames for MEDSEP Blood Bag # 791 05, 0791 05U &8 0346 O (7 Canisters)
SDP
Catalog No. 5000662016
Encompass_Preferred
Canister Frames are constructed of corrosion resistant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Frame for 7 MEDSEP CBBC V1 Aluminum Cord Blood Bag Canisters Blood Bag Number: 791 OS 791 OSU 8 0346 O Dimensions (D x H x L): 3/14 x 26 7/8 x 9/16″ 82 x 683 x 15mm Canister: CBBC V1 Platform Dividers P20 CBBC V P24 CBBC V P31 CBBC V P39 CBBC V
Catalog No. 50-006-62016 Supplier Alkali Scientific Supplier No. FCCBBCV17
Canister Frames are constructed of corrosion resistant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Frame for 7 MEDSEP CBBC V1 Aluminum Cord Blood Bag Canisters Blood Bag Number: 791 OS 791 OSU 8 0346 O Dimensions (D x H x L): 3/14 x 26 7/8 x 9/16″ 82 x 683 x 15mm Canister: CBBC V1 Platform Dividers P20 CBBC V P24 CBBC V P31 CBBC V P39 CBBC V