Learn More
Alkali Scientific Stainless Steel Canister Frames for STERICON Blood Bag # RCM 91D (5 Canisters)
SDP
Catalog No. 5000662358
Encompass_Preferred
Canister Frames are constructed of corrosion resistant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Frame for 5 STERICON C 91D Aluminum Canisters Blood Bag Number: RCM 91D Dimensions (D x H x L): 10 3/16 x 28 9/16 x 9/16″ 259 x 726 x 15 mm Canister: C 91D Platform Dividers P20 91D P24 91D P31 91D P39 91D
Catalog No. 50-006-62358 Supplier Alkali Scientific Supplier No. FC91D5
Canister Frames are constructed of corrosion resistant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Frame for 5 STERICON C 91D Aluminum Canisters Blood Bag Number: RCM 91D Dimensions (D x H x L): 10 3/16 x 28 9/16 x 9/16″ 259 x 726 x 15 mm Canister: C 91D Platform Dividers P20 91D P24 91D P31 91D P39 91D