Learn More
METTLER TOLEDO™ Stainless Steel Capsule Adapter Bottles
Catalog No. 01676976 Shop All METTLER TOLEDO Products
Change view
Click to view available options
METTLER TOLEDO™ Adapter Bottle
Catalog No.
01-676-976 METTLER TOLEDO™ Adapter Bottle
1 options
Catalog No. 01-676-976 Supplier METTLER TOLEDO™ Supplier No. 30459925
Only null left
Add to Cart
Add to Cart
Adapter accessories are made with conductive materials to reduce electrostatic charge build up. Our adapters load capsules, vials, bottles, or tubes in precisely the right position and secure them tightly in place for highly accurate dosing.
  • Secures vessel in precise position
  • Compatible with Q3 Sample Changer
  • Conductive materials to reduce electrostatic charge build-up
  • Highly accurate and safe filling
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Excellence XPR Automatic Balance
Size (Metric) 44.5 x 15 mm
Type Adapter Bottle