Learn More
MSC Albion Cam Brake, Stainless Steel, Short Pedal, 1 ea
Catalog No. 5000512163
Encompass
This cam brake accessory is designed to enhance the functionality and control of industrial casters. Constructed from durable stainless steel, this component provides reliable locking capabilities for various material handling applications.
  • Constructed from stainless steel for enhanced durability
  • Functions as a cam brake for caster systems
  • Compatible with 12, 16, 18, 60, 61, 62, 71, and 72 series casters
  • Designed for use with 4 in. and 5 in. caster models
Catalog No. 50-005-12163 Supplier MSC Supplier No. 63280192
This cam brake accessory is designed to enhance the functionality and control of industrial casters. Constructed from durable stainless steel, this component provides reliable locking capabilities for various material handling applications.
  • Constructed from stainless steel for enhanced durability
  • Functions as a cam brake for caster systems
  • Compatible with 12, 16, 18, 60, 61, 62, 71, and 72 series casters
  • Designed for use with 4 in. and 5 in. caster models