Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Chest Freezer Rack for MATRIX Boxes, FluidX Boxes & Micronic Racks, 6 Box Capacity
SDP
Catalog No. 503029233
Encompass_Preferred
For Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 2 3/16″- 2 7/16″ (54.8-61.8mm) The CFMX-6-2 vertical freezer racks are designed to safely store a total of (6) 96 Well MATRIX Boxes, FluidX Boxes or Micronic Racks in a laboratory chest freezer or ln2 freezer. These racks are made out of corrosion resistant stainless steel and include locking rods and handles. Boxes are not included with purchase. Dimensions H x D x W: 15 3/16 x 3 5/8 x 5 1/2inch (260 x 92 x 140mm)Compartment Dimensions h x d x w: 2 1/2 x 3 7/16 x 51/8 inch (62.5 x 88 x 130mm)
Catalog No. 50-302-9233 Supplier Alkali Scientific Supplier No. CFMX62
For Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 2 3/16″- 2 7/16″ (54.8-61.8mm) The CFMX-6-2 vertical freezer racks are designed to safely store a total of (6) 96 Well MATRIX Boxes, FluidX Boxes or Micronic Racks in a laboratory chest freezer or ln2 freezer. These racks are made out of corrosion resistant stainless steel and include locking rods and handles. Boxes are not included with purchase. Dimensions H x D x W: 15 3/16 x 3 5/8 x 5 1/2inch (260 x 92 x 140mm)Compartment Dimensions h x d x w: 2 1/2 x 3 7/16 x 51/8 inch (62.5 x 88 x 130mm)