Learn More
Troemner™ Stainless Steel Class 3 Weight
Catalog No. 02218235 Shop All Troemner Products
Change view
Click to view available options
Calibration Weight; Weight: 100 mg
Catalog No.
02-218-235 Calibration Weight; Weight: 100 mg
1 options
Catalog No. 02-218-235 Supplier Troemner™ Supplier No. 80858448
Add to Cart
Add to Cart
Troemener Individual Calibration Weights are Adjusted to Meet or Exceed ASTM Class 1-4 Tolerances
  • Troemner ASTM Class 1-4 offer high accuracy with tight tolerances and uncertainties of less than 1/4 of the tolerance. Weight surfaces meet or exceed ASTM and have a highly polished finish.
  • Available in high grade stainless steels including Troemner Alloy 8 which possesses closely controlled density, extremely low magnetic properties, good stability, and resistance to corrosion.
  • Available with certificate options traceable to NIST including ISO/IEC 17025 Accredited Certificate or Traceable Certificate to aid in meeting SOP and regulatory requirements.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Calibration Mass
Certifications/Compliance No Certificate
Material Stainless Steel (304)
Case Material Polycarbonate
Weight (Metric) 100 mg
Shape Flat