Learn More
Troemner™ Stainless Steel Class 3 Weight with ISO/IEC 17025 Accredited Certificate
Shop All Troemner ProductsDescription
- Troemner ASTM Class 1-4 offer high accuracy with tight tolerances and uncertainties of less than 1/4 of the tolerance. Weight surfaces meet or exceed ASTM and have a highly polished finish.
- Available in high grade stainless steels including Troemner Alloy 8 which possesses closely controlled density, extremely low magnetic properties, good stability, and resistance to corrosion.
- Available with certificate options traceable to NIST including ISO/IEC 17025 Accredited Certificate or Traceable Certificate to aid in meeting SOP and regulatory requirements.
Specifications
Specifications
| Product Type | Calibration Mass |
| Certifications/Compliance | ISO/IEC 17025 Accredited Certificate |
| Material | Stainless Steel (304) |
| Case Material | Polycarbonate |
| Weight (Metric) | 200 mg |
| Shape | Flat |