Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ Stainless Steel Class 7 Slotted Weight Set

Catalog No. 19216381 Shop All Troemner Products
Catalog No. 19-216-381 Supplier Troemner™ Supplier No. 30390729
Add to Cart
Add to Cart

Manufactured from stainless steel. The Troemner™ Stainless Steel Class 7 Slotted Weight Set is used for rough weighing operations in physical and chemical laboratories.

  • One-piece construction
  • Case sold separately
  • Rack storage
TRUSTED_SUSTAINABILITY

Specifications

Product Type Slotted Weight Set with No Certificate
Classification Class 7
Material Weights above 5 g are 303 Stainless Steel. Weights 5 g and below are 3003-H14 Aluminum.
Density Weights above 5 g 7.85 g/cm³ at 20°C. Weights 5 g and below are 2.7 g/cm³ at 20°C.
Includes 500 g, (5) 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.