Learn More
Troemner™ Stainless Steel Class 7 Weight Set
Catalog No. 19216364 Shop All Troemner Products
Catalog No. 19-216-364 Supplier Troemner™ Supplier No. 30390964
Add to Cart
Add to Cart
Manufactured from stainless steel. The Troemner™ Stainless Steel Class 7 Weight Set is used for rough weighing operations in physical and chemical laboratories.
  • One- and two-piece construction, depending on weight
  • Includes case with latch
  • Made in the USA
TRUSTED_SUSTAINABILITY
Specifications
Product Type Weight Set with No Certificate
Classification Class 7
Material Stainless Steel
Case Material Plastic
Height (English) Case 1.7 in.
Includes 500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg