Learn More
Troemner™ Stainless Steel Class 7 Weight Set
Catalog No. 19216369 Shop All Troemner Products
Catalog No. 19-216-369 Supplier Troemner™ Supplier No. 30390969
Add to Cart
Add to Cart
Manufactured from stainless steel. The Troemner™ Stainless Steel Class 7 Weight Set is used for rough weighing operations in physical and chemical laboratories.
  • One- and two-piece construction, depending on weight
  • Includes case with latch
  • Made in the USA
TRUSTED_SUSTAINABILITY
Specifications
Product Type Weight Set with No Certificate
Certifications/Compliance Class 7
Material Stainless Steel
Case Material Plastic
Height (English) Case 0.836
Includes 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g