Learn More
Troemner™ Stainless Steel Class 7 Weight Set
Catalog No. 19216370 Shop All Troemner Products
Catalog No. 19-216-370 Supplier Troemner™ Supplier No. 30390972
Add to Cart
Add to Cart
Manufactured from stainless steel. The Troemner™ Stainless Steel Class 7 Weight Set is used for rough weighing operations in physical and chemical laboratories.
  • One- and two-piece construction, depending on weight
  • Includes certificate of accuracy
  • Includes case with latch
  • Made in the USA
TRUSTED_SUSTAINABILITY
Specifications
Product Type Weight Set with No Certificate
Classification Class 7
Material Stainless Steel
Case Material Plastic
Includes 50g, (2) 20g, 10g, 5g, (2) 2g, 1g