Learn More
MSC ZEP Stainless Steel Cleaner and Polish, Aerosol, 14 oz, 4/Case
Catalog No. 502868803
Encompass
Zep Stainless Steel Cleaner and Polish is a professional-grade formula designed to clean and protect stainless steel, chrome, and aluminum surfaces. It effectively removes fingerprints, watermarks, and dirt, leaving surfaces gleaming without residue. Suitable for use on various appliances and helps prevent rust, corrosion, and discoloration.
  • Cleans and polishes in one step
  • Prevents rust and corrosion
  • Fast-drying, streak-free formula
  • Removes smudges, fingerprints, and watermarks
  • Made in the USA
Catalog No. 50-286-8803 Supplier MSC Supplier No. 17641531
Zep Stainless Steel Cleaner and Polish is a professional-grade formula designed to clean and protect stainless steel, chrome, and aluminum surfaces. It effectively removes fingerprints, watermarks, and dirt, leaving surfaces gleaming without residue. Suitable for use on various appliances and helps prevent rust, corrosion, and discoloration.
  • Cleans and polishes in one step
  • Prevents rust and corrosion
  • Fast-drying, streak-free formula
  • Removes smudges, fingerprints, and watermarks
  • Made in the USA