Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Stainless Steel Cleaner & Protector with Scotchgard™

Catalog No. 17623932 Shop All 3M Products
Change view
Click to view available options
Volume (English):
1 qt.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
17-623-932 1 qt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-623-932 Supplier 3M™ Supplier No. 7100082687
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

3M™ Stainless Steel Cleaner and Protector with Scotchgard™ leaves an invisible layer of protection on stainless steel surfaces making them easier to clean. This water based, oil free formula allows for quick drying streak free spot cleaning to reduce labor and lower product consumption.

  • Clean with less effort
  • Allows spot cleaning
  • Streak-free shine
  • NSF A7 certified for non-direct food contact surfaces
  • USDA certified biobased product
  • Dries quickly
  • Non-corrosive
  • Non-flammable
TRUSTED_SUSTAINABILITY

Specifications

Product Type Stainless Steel Cleaner
Form Liquid
Certifications/Compliance NSF A7
Concentration RTU
Container Type Flip-Top Cap
For Use With (Application) Stainless Steel Cleaning
Volume (English) 1 qt.
Volume (Metric) 946.4 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.