Learn More
TA Instruments Stainless Steel, Composite HB, 25 mm, NST, Upper Plate with Composite Heat Break, for Use with Lower Peltier Plate, Smart Swap 2 Geometry, HR x0 Instruments Only
Catalog No. 5000580805 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, Stainless Steel, Composite HB, 25 mm, NST is an upper plate with composite heat break for use with the lower Peltier plate. It has Smart Swap 2 geometry and is for use with HR X0 instruments only.
Catalog No. 50-005-80805 Supplier TA Instruments Supplier No. 512250.946
Waters TA Instruments HR X0, Stainless Steel, Composite HB, 25 mm, NST is an upper plate with composite heat break for use with the lower Peltier plate. It has Smart Swap 2 geometry and is for use with HR X0 instruments only.