Learn More
TA Instruments Stainless Steel, Composite HB, 40 mm, 2 Degree Cone, Upper Cone for Use with Lower Peltier Plate, Smart Swap 2 Geometry, HR x0 Instruments Only
Catalog No. 5000580773 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, Stainless Steel, Composite HB, 40 mm, 2 Degree Cone is an upper cone for use with the lower Peltier plate. It has Smart Swap 2 geometry and is for use with HR X0 instruments only.
Catalog No. 50-005-80773 Supplier TA Instruments Supplier No. 512406.945
Waters TA Instruments HR X0, Stainless Steel, Composite HB, 40 mm, 2 Degree Cone is an upper cone for use with the lower Peltier plate. It has Smart Swap 2 geometry and is for use with HR X0 instruments only.