Learn More
TA Instruments Stainless Steel, Conical DIN Rotor, Radius 14 mm, Rotor Height 42 mm, HR X0, Smart Swap 2
Catalog No. 5000580702 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, Smart Swap 2, Stainless Steel, Conical DIN Rotor has a radius of 14 mm and a rotor height of 42 mm.
Catalog No. 50-005-80702 Supplier TA Instruments Supplier No. 546012.941
Waters TA Instruments HR X0, Smart Swap 2, Stainless Steel, Conical DIN Rotor has a radius of 14 mm and a rotor height of 42 mm.