Learn More
TA Instruments Stainless Steel, Conical DIN Rotor, Sandblasted, Radius 14 mm, Rotor Height 42 mm, Surface Finish 80-84 RMS, Ra 1.8-2.0 Microns, HR X0
Catalog No. 5000580644 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, Stainless Steel, Conical DIN Rotor, Sandblasted has a radius of 14 mm and rotor height of 42 mm. The surface finish is 80-84 RMS in micro inches or Ra value of 1.8 to 2.0 microns.
Catalog No. 50-005-80644 Supplier TA Instruments Supplier No. 546009.941
Waters TA Instruments HR X0, Stainless Steel, Conical DIN Rotor, Sandblasted has a radius of 14 mm and rotor height of 42 mm. The surface finish is 80-84 RMS in micro inches or Ra value of 1.8 to 2.0 microns.