Learn More
TA Instruments Stainless Steel, Conical Rotor, Radius 7.5 mm, Rotor Height 42 mm, for Crumb-Rubber Modified Asphalt, HR X0
Catalog No. 5000580760 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, Stainless Steel, Conical Rotor has a radius of 7.5 mm and rotor height of 42 mm. It is appropriate for measurements of crumb-rubber modified asphalt.
Catalog No. 50-005-80760 Supplier TA Instruments Supplier No. 545043.941
Waters TA Instruments HR X0, Stainless Steel, Conical Rotor has a radius of 7.5 mm and rotor height of 42 mm. It is appropriate for measurements of crumb-rubber modified asphalt.