Learn More
AVESTIN INC Avestin Stainless Steel Cooling Coil
Catalog No. NC1802080
Encompass
316 stainless steel Avestin allowing for simple non stringent temperature control, cooling or heating of the sample. Offers pronounced heat loss and effective cooling when immersed in an ice or water bath. N.B - Only suitable for use with homogenisigin pressures of up to 20,000psi.
Catalog No. NC1802080 Supplier AVESTIN INC Supplier No. C5HECOIL1
316 stainless steel Avestin allowing for simple non stringent temperature control, cooling or heating of the sample. Offers pronounced heat loss and effective cooling when immersed in an ice or water bath. N.B - Only suitable for use with homogenisigin pressures of up to 20,000psi.