Learn More
Analytical Sales & Services Stainless Steel Cross for 1/16" Tubing
Catalog No. 5000806586
Encompass
Advantage SS Cross for 1/16" Tubing 0.25mm Thru-Hole\n\nQty/Pkg: EACH
Catalog No. 50-008-06586 Supplier Analytical Sales & Services Supplier No. 1603
Advantage SS Cross for 1/16" Tubing 0.25mm Thru-Hole\n\nQty/Pkg: EACH