Learn More
TA Instruments Stainless Steel, Cross Hatched, 20 mm Plate, NST, HR X0
Catalog No. 5000580657 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0 Stainless Steel Cross Hatched Upper Plate (20 mm, NST) is for use with lower Peltier plate. Smart Swap 2 geometry for use with HR X0 instruments only. Cross hatching 1 mm deep, 1 mm peak-to-peak.
Catalog No. 50-005-80657 Supplier TA Instruments Supplier No. 517200.946
Waters TA Instruments HR X0 Stainless Steel Cross Hatched Upper Plate (20 mm, NST) is for use with lower Peltier plate. Smart Swap 2 geometry for use with HR X0 instruments only. Cross hatching 1 mm deep, 1 mm peak-to-peak.