Learn More
TA Instruments Stainless Steel, Cross Hatched, 20 mm Plate, Smart Swap 2 Geometry, HR x0 Instruments Only
Supplier:  TA Instruments 517200.945
Encompass_Preferred
HR X0 Stainless Steel Cross Hatched Plate (20 mm) is an upper plate for use with lower Peltier plates. It features Smart Swap 2 geometry and 1 mm deep, 1 mm peak-to-peak cross hatching for improved sample retention and measurement consistency on HR X0 instruments.
Catalog No. 50-005-80854