Learn More
TA Instruments Stainless Steel, Cross Hatched, 20 mm, Quick Change Plate, HR X0, DHR, ARES-G2, APS
Catalog No. 5000580871 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, DHR, ARES-G2, APS Stainless Steel Cross Hatched Quick Change Plate (20 mm) features 1 mm deep, 1 mm peak-to-peak cross hatching. Compatible with Advanced Peltier Plates and ARES-G2 APS systems, ensuring secure placement and consistent sample testing.
Catalog No. 50-005-80871 Supplier TA Instruments Supplier No. 402492.901
Waters TA Instruments HR X0, DHR, ARES-G2, APS Stainless Steel Cross Hatched Quick Change Plate (20 mm) features 1 mm deep, 1 mm peak-to-peak cross hatching. Compatible with Advanced Peltier Plates and ARES-G2 APS systems, ensuring secure placement and consistent sample testing.