Learn More
TA Instruments Stainless Steel, Cross Hatched, 40 mm, Quick Change Plate, HR X0, DHR, ARES-G2, APS
Catalog No. 5000580750 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, DHR, ARES-G2, APS Stainless Steel Cross Hatched Quick Change Plate (40 mm) features 1 mm deep, 1 mm peak-to-peak cross hatching. Compatible with Advanced Peltier Plates and ARES-G2 APS Standard Flat Plate, providing secure sample placement and reproducible measurement.
Catalog No. 50-005-80750 Supplier TA Instruments Supplier No. 402629.901
Waters TA Instruments HR X0, DHR, ARES-G2, APS Stainless Steel Cross Hatched Quick Change Plate (40 mm) features 1 mm deep, 1 mm peak-to-peak cross hatching. Compatible with Advanced Peltier Plates and ARES-G2 APS Standard Flat Plate, providing secure sample placement and reproducible measurement.