Learn More
TA Instruments Stainless Steel, Cross Hatched, 8 mm, Quick Change Plate, HR X0, DHR, ARES-G2, APS
Catalog No. 5000580847 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, DHR, ARES-G2, APS Stainless Steel Cross Hatched Quick Change Plate (8 mm) features 1 mm deep, 1 mm peak-to-peak cross hatching. Designed for Advanced Peltier Plates and ARES-G2 APS systems to improve sample grip, consistent placement, and reproducible rheological testing.
Catalog No. 50-005-80847 Supplier TA Instruments Supplier No. 402627.901
Waters TA Instruments HR X0, DHR, ARES-G2, APS Stainless Steel Cross Hatched Quick Change Plate (8 mm) features 1 mm deep, 1 mm peak-to-peak cross hatching. Designed for Advanced Peltier Plates and ARES-G2 APS systems to improve sample grip, consistent placement, and reproducible rheological testing.