Learn More
OHAUS Stainless Steel Defender 5000 Series Floor Scale
Catalog No. 01920178 Shop All OHAUS Products
Change view
Click to view available options
Model:
i-DF52XW2500C1L
i-DF52XW2500C1R
i-DF52XW5000C1L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
01-920-178 i-DF52XW5000C1L
01-920-179 i-DF52XW2500C1L
01-920-182 i-DF52XW2500C1R
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-920-178 Supplier OHAUS Supplier No. 30837701
Add to Cart
Add to Cart
Request a Quote
Defender 5000 Series Floor Scales meet advanced weighing needs for industrial and commercial use. Their high-quality construction is designed to minimize flexing under heavy loads for years of dependable performance. The indicator features software for counting, checkweighing, accumulation, display hold and other modes.
  • 304 stainless steel, flat surface decks with 4 IP68 load cells, fixed top plate (5 mm/0.196 in. thick) with structural bracing for stiff construction.
  • Platforms available in sizes of 3 x 3 ft. and 4 x 4 ft. and capacities from 2,500 lb. to 5,000 lb. with NTEP certified 5,000e performance, allowing greater flexibility for your weighing needs.
  • Equipped with a new TD52 series indicator featuring an easy-to-use keypad, large backlit display, and plain text on-screen prompts to guide you through operation.
TRUSTED_SUSTAINABILITY
Specifications
Capacity 2500 kg
Type Scale
Display Type LCD
Readability 0.5 kg
Legal for Trade Class III
Model i-DF52XW5000C1L