Learn More
Hach Company Stainless Steel Digital Contacting Conductivity Sensor (k=0.5) for Low-Conductivity Boiler and Condensate Monitoring, includes digital gateway
Catalog No. 5000207544 Shop All Hach Company Products
Encompass
Digital contacting conductivity sensor housed in a 316 stainless steel body for low-conductivity boiler and condensate monitoring. The sensor provides wide-range conductivity measurement, built-in temperature sensing, and is supplied with a digital gateway and extension cable for integration with digital controllers.
  • Nominal cell constant 0.5 cm⁻¹ for low-conductivity applications.
  • Measures from ultra-pure water (0.057 μS/cm) up to 200,000 μS/cm.
  • Accuracy ±2% of reading above 200 μS/cm and Pt1000 RTD temperature accuracy ±0.1 °C.
  • 3/4 in male NPT thread for standard installation and 72.39 mm immersion depth.
  • Supplied with a digital gateway, fixed cable, and a digital extension cable for easy integration.
  • Operating range -20 to 200 °C and pressure rating up to 20.7 bar.
  • Repeatability and sensitivity of 0.5% support reliable process control.
Catalog No. 50-002-07544 Supplier Hach Company Supplier No. D3444B8
Digital contacting conductivity sensor housed in a 316 stainless steel body for low-conductivity boiler and condensate monitoring. The sensor provides wide-range conductivity measurement, built-in temperature sensing, and is supplied with a digital gateway and extension cable for integration with digital controllers.
  • Nominal cell constant 0.5 cm⁻¹ for low-conductivity applications.
  • Measures from ultra-pure water (0.057 μS/cm) up to 200,000 μS/cm.
  • Accuracy ±2% of reading above 200 μS/cm and Pt1000 RTD temperature accuracy ±0.1 °C.
  • 3/4 in male NPT thread for standard installation and 72.39 mm immersion depth.
  • Supplied with a digital gateway, fixed cable, and a digital extension cable for easy integration.
  • Operating range -20 to 200 °C and pressure rating up to 20.7 bar.
  • Repeatability and sensitivity of 0.5% support reliable process control.