Learn More
TA Instruments Stainless Steel, Disposable Plate Shaft for UV Accessories, for Use with UV-Curing Options and Disposable Stepped Peltier Plate, Not for Use with Environmental Test Chamber ETC, HR x0
Catalog No. 5000580839 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, Stainless Steel, Disposable Plate Shaft for UV Accessories is for use with UV-curing options and the disposable stepped Peltier plate. It is not for use with the Environmental Test Chamber ETC.
Catalog No. 50-005-80839 Supplier TA Instruments Supplier No. 546320.941
Waters TA Instruments HR X0, Stainless Steel, Disposable Plate Shaft for UV Accessories is for use with UV-curing options and the disposable stepped Peltier plate. It is not for use with the Environmental Test Chamber ETC.