Learn More
Mopec Stainless Steel Dissecting Table
SDP
Catalog No. 22444313 Shop All Mopec Products
Change view
Click to view available options
Description:
Collapsable 12-inch legs
Standard legs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
22-444-313 Standard legs
22-444-321 Collapsable 12-inch legs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-444-313 Supplier Mopec Supplier No. BC006
Add to Cart
Add to Cart
Mopec Stainless Steel Dissecting Table can accommodate large or small standard dissection boards
  • Work surface is tapered toward a centrally located drainage hole
  • 3-inch drain hose (provided) can be connected to a fitting on the underside of the tray
  • Sturdy legs (optional collapsible) can be set on any counter or autopsy table
Specifications
Product Type Dissecting Table
Material Stainless Steel
Description Standard legs
For Use With (Equipment) Dissecting Boards
Length (English) 25 in.
Width (English) 18 in.