Learn More
TA Instruments Stainless Steel Double Gap Concentric Cylinder Cup, Diameter 30.4 mm, Outer Radius 18.5 mm, for HR X0, DHR and AR-Series Peltier Jacket
Catalog No. 5000580662 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Stainless Steel Double Gap Concentric Cylinder Cup has a diameter of 30.4 mm and outer radius of 18.5 mm. It is for use with HR X0, DHR, and AR-Series Peltier Jacket.
Catalog No. 50-005-80662 Supplier TA Instruments Supplier No. 546045.901
Waters TA Instruments Stainless Steel Double Gap Concentric Cylinder Cup has a diameter of 30.4 mm and outer radius of 18.5 mm. It is for use with HR X0, DHR, and AR-Series Peltier Jacket.