Learn More
TA Instruments Stainless Steel, Double Gap Rotor, Inner Radius 16 mm, Outer Radius 17.5 mm, Height 53 mm, HR X0, Smart Swap 2
Catalog No. 5000580837 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, Smart Swap 2, Stainless Steel, Double Gap Rotor has an inner radius of 16 mm, outer radius of 17.5 mm, and height of 53 mm.
Catalog No. 50-005-80837 Supplier TA Instruments Supplier No. 546048.941
Waters TA Instruments HR X0, Smart Swap 2, Stainless Steel, Double Gap Rotor has an inner radius of 16 mm, outer radius of 17.5 mm, and height of 53 mm.