Learn More
TA Instruments Stainless Steel, Double Gap Rotor, Inner Radius 20.38 mm, Outer Radius 21.96 mm, Height 59.5 mm, HR X0, Smart Swap 2
Catalog No. 5000580703 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, Smart Swap 2, Stainless Steel, Double Gap Rotor has an inner radius of 20.38 mm, outer radius of 21.96 mm, and height of 59.5 mm.
Catalog No. 50-005-80703 Supplier TA Instruments Supplier No. 546022.941
Waters TA Instruments HR X0, Smart Swap 2, Stainless Steel, Double Gap Rotor has an inner radius of 20.38 mm, outer radius of 21.96 mm, and height of 59.5 mm.