Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ Stainless Steel Electronic Balance Calibration Weights Sets

Catalog No. 022147T Shop All Troemner Products
Change view
Click to view available options
Certifications/Compliance:
NVLAP, ISO9001
No certificate
Traceable certificate, ISO9001
Classification:
Class 1
Class 4
UltraClass
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance Classification
02-214-7T Traceable certificate, ISO9001 UltraClass
02-216-022 NVLAP, ISO9001 Class 1
02-214-8T NVLAP, ISO9001 UltraClass
02-214-6T No certificate UltraClass
02-216-024 No certificate Class 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02-214-7T Supplier Troemner™ Supplier No. 80860834
Add to Cart
Add to Cart

100g to 1kg

  • 2 piece constructions
  • Weights below 1 kg are straight cylinders
  • 1kg weights have groove neare thet top for easy handling
  • Weight set configurations: 1kg, 500g, 300g, 200g, 100g
  • Total capacity of weight set: 2100g
  • UltraClass: With weight tolerances 50% greater than ANSI/ASTM E617 Class 1 tolerances
  • ANSI/ASTM Class 1: Can be used as a reference standard in calibrating other weights and is appropriate for calibrating high precision analytical balances with a readability as low as 0.1mg to 0.01mg
  • ANSI/ASTM Class 2: Appropriate for calibrating high-precision top loading balances with a readability as low as 0.01g to 0.001g
  • ANSI/ASTM Class 3: Appropriate for calibrating balances with moderate precision, with a readability as low as 0.1g to 0.01g
  • ANSI/ASTM Class 4: For calibration of semi-analytical balances and for student use

Order Info

Supplied with impact-resistant polypropylene case.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Electronic Balance Calibration Weight
Certifications/Compliance Traceable certificate, ISO9001
Classification UltraClass
Material Stainless Steel
Weight (Metric) 100 g to 1 kg
Density 7.95g/cm3 at 20°C
Mass 100 g to 1 kg
Includes Cylindrical weight: 1kg, 500g, 300g, 200g, 100g, Plastic case, white Cotton glove
Quantity 5/Set
Packaging Quantity Multiple
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.