Learn More
Analytical Sales & Services Stainless Steel Ferrules for 1/8'' OD Tubing
Catalog No. 5000804319
Encompass
Ferrule for 1/8” OD Tubing, 1/4-28 Port
Catalog No. 50-008-04319 Supplier Analytical Sales & Services Supplier No. 52236X
Ferrule for 1/8” OD Tubing, 1/4-28 Port