Learn More
Description
- Constructed of stainless steel
- For use with flasks
- Designed to securely hold flask in place
Specifications
Specifications
| Type | Flask Clamp |
| Material | Stainless Steel |
| For Use With (Equipment) | 250 mL Flask |
| Description | Flask Clamp, 250 mL |
| Color | Silver |
| Jaw Type | Four Interlocking |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.