Learn More
SOMNI Scientific™ Stainless Steel Flowmeter Delivery System
SDP
Catalog No. 04777112 Shop All Somni Scientific Products
Change view
Click to view available options
For Use With (Application):
Research Animal Inhalant Anesthesia
Catalog No. For Use With (Application)
04-777-112 Research Animal Inhalant Anesthesia
1 options
Catalog No. 04-777-112 Supplier SOMNI Scientific™ Supplier No. DS14004
Add to Cart
Add to Cart
Creates the ability for one system to provide adjustable flowrates to multiple stations
  • Easily adapted to any existing anesthesia system
  • Upgrade from the standard stopcock design
  • Complete with color coded delivery lines and adapters (2 Station to Mount on to TT2)
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia
For Use With (Equipment) RM-17004, TT-17000, TT-17001, TT-17002
Material Aluminum, Stainless Steel, Glass, Plastic
Length (Metric) 7.62 cm
Width (Metric) 5.08 cm
Includes Color-coded delivery lines and adapters
For Veterinary Use Only