Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Stainless Steel Foot Rest
GREENER_CHOICE

Catalog No. 15050338 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W x H):
21 x 11 x 13 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
15-050-338 21 x 11 x 13 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 15-050-338
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stainless Steel Foot Rests are constructed from heavy gauge Type 304 stainless steel with an all-welded construction.

  • Stainless Steel Foot Rests are constructed from heavy gauge Type 304 stainless steel with an all-welded construction.
  • The frame is 1 in. square tube with slotted uprights.
  • The platform is at an ergonomic angle of 12O and is adjustable on 1.5 in. centers ranging from 4.5 in. to 9.75 in.
  • It includes non-slip rubber feet and is ideal for use in cleanrooms, labs healthcare facilities; any place where long periods of sitting is required.

Specifications

Product Type Foot Rest
Dimensions (L x W x H) 21 x 11 x 13 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.