Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Freezer Rack for Chest Freezers or LN2 Tanks for 15mL & 50mL Tube Boxes, 4 Box Capacity
SDP
Supplier:  Alkali Scientific CFLB4
Encompass_Preferred
Easily place and retrieve racks using sturdy dual handles Included locking rod securely holds boxes in place Sturdy corrosion-resistant stainless-steel construction for long-term use Easily place and retrieve racks using sturdy dual handles Included locking rod securely holds boxes in place. Sturdy corrosion-resistant stainless-steel construction for long-term use Racks feature sturdy handles for easy access to your samples in vertical/chest freezers.  Dimensions H x D x W: 20 3/8 x 6 x 6 3/16 Inch ( 517 x 153 x 157mm)Compartment Dimensions: 5 x 5 7/8 x 5 7/8 inch (126.8 x 149 x149mm)   **Lock device is ordered separately.
Catalog No. 50-302-9201