Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Freezer Racks for 3" Boxes, 3 x 3 Configuration, 9 Box Capacity
SDP
Catalog No. 503026398
Encompass_Preferred
Constructed of corrosion resistant stainless steel Racks fit standard 3" cardboard, plastic, or metal boxes Security lock devices available for each rack Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-hand doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions (DxWxH): 16 7/8 x 5 1/2 x 9 3/8 inches (429 x 140 x 238mm)Compartment Dimensions: 5 3/8 x 5 7/16 x 3 1/16 inches (137 x 140 x 78mm)
Catalog No. 50-302-6398 Supplier Alkali Scientific Supplier No. UFR333
Constructed of corrosion resistant stainless steel Racks fit standard 3" cardboard, plastic, or metal boxes Security lock devices available for each rack Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-hand doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions (DxWxH): 16 7/8 x 5 1/2 x 9 3/8 inches (429 x 140 x 238mm)Compartment Dimensions: 5 3/8 x 5 7/16 x 3 1/16 inches (137 x 140 x 78mm)