Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Freezer Racks for 2" Boxes, Vertical Configuration, with Spring Clip, 9 Box Capacity
SDP
Catalog No. 503026411
Encompass_Preferred
Our Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers. Vertical Racks for Chest Freezers and Liquid Nitrogen Tanks are made of corrosion resistant stainless steel.  Racks are made for standard 2" boxes.  All racks include stainless steel locking rods.  Security lock devices are also available for each rack though ordered separately. Features: Constructed of corrosion resistant stainless steel Racks fit standard 2" cardboard, plastic, or metal boxes Security lock devices available for each rack Dimensions H x W x D: 20 1/16 x 5 5/8 x 5 1/2 inch (510 x 143 x 140mm)Compartment Dimensions h x w x d:2 3/16 x 5 5/16 x 5 5/16 inch ( 55 x 134.5 x 135mm) **Lock device is ordered separately.
Catalog No. 50-302-6411 Supplier Alkali Scientific Supplier No. CF92S
Our Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers. Vertical Racks for Chest Freezers and Liquid Nitrogen Tanks are made of corrosion resistant stainless steel.  Racks are made for standard 2" boxes.  All racks include stainless steel locking rods.  Security lock devices are also available for each rack though ordered separately. Features: Constructed of corrosion resistant stainless steel Racks fit standard 2" cardboard, plastic, or metal boxes Security lock devices available for each rack Dimensions H x W x D: 20 1/16 x 5 5/8 x 5 1/2 inch (510 x 143 x 140mm)Compartment Dimensions h x w x d:2 3/16 x 5 5/16 x 5 5/16 inch ( 55 x 134.5 x 135mm) **Lock device is ordered separately.