Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Freezer Racks for 3.75" Boxes, 4 x 3 Configuration, 12 Box Capacity
SDP
Catalog No. 503027910
Encompass_Preferred
Constructed of corrosion resistant stainless steel Racks fit standard 3.75" cardboard, plastic, or metal boxes Security lock devices available for each rack 4 Long x 3 Tall= Holds 12 Boxes  Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-handed doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions L x H x W: 22 1/8 x 12 1/16 x 5 1/2 inch (562 x 307  x 140mm)Compartment Size L x H x W: 5 3/8 x 3 15/16 x 5 3/8 inch (136 x 100 x137mm)
Catalog No. 50-302-7910 Supplier Alkali Scientific Supplier No. UFR43375
Constructed of corrosion resistant stainless steel Racks fit standard 3.75" cardboard, plastic, or metal boxes Security lock devices available for each rack 4 Long x 3 Tall= Holds 12 Boxes  Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-handed doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions L x H x W: 22 1/8 x 12 1/16 x 5 1/2 inch (562 x 307  x 140mm)Compartment Size L x H x W: 5 3/8 x 3 15/16 x 5 3/8 inch (136 x 100 x137mm)