Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Freezer Racks for Large 3.75" Boxes, 4 x 3 Configuration, 12 Box Capacity
SDP
Catalog No. 503027911
Encompass_Preferred
Constructed of corrosion resistant stainless steel Racks fit standard 3.75" cardboard, plastic, or metal boxes Security lock devices available for each rack 4 Long x 3 Tall= Holds 12 Boxes  Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-handed doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions L x H x W: 24 5/16 x 12 1/16 x 5 15/16 (315.5 x 307 x 151mm)Compartment Dimensions: 5 7/8 x 4 x 5 13/16 inch (149 x 101 x 148mm)
Catalog No. 50-302-7911 Supplier Alkali Scientific Supplier No. UFR43L375
Constructed of corrosion resistant stainless steel Racks fit standard 3.75" cardboard, plastic, or metal boxes Security lock devices available for each rack 4 Long x 3 Tall= Holds 12 Boxes  Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-handed doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions L x H x W: 24 5/16 x 12 1/16 x 5 15/16 (315.5 x 307 x 151mm)Compartment Dimensions: 5 7/8 x 4 x 5 13/16 inch (149 x 101 x 148mm)