Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Freezer Racks with Drawers, 5 x 6 Configuration, 30 Box Capacity, for 2" Boxes
SDP
Catalog No. 503028001
Encompass_Preferred
Constructed of corrosion resistant stainless steel Racks fit standard 2" cardboard, plastic, or metal boxes Security lock devices available for each rack Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-hand doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Upright Freezer Drawer racks offer a more organized system for storage and retrieval of samples. Drawer racks facilitate both access to and storage of samples without having to remove the whole rack from the freezer. Dimensions (D x H x W): 27  x 14 1/8 x 5 1/2 inches (687 x 359 x 140mm)Compartment Dimensions: 5 3/8  x 2 1/16x 5 1/4inches (137.4 x 52 x 133mm)
Catalog No. 50-302-8001 Supplier Alkali Scientific Supplier No. UFDR562
Constructed of corrosion resistant stainless steel Racks fit standard 2" cardboard, plastic, or metal boxes Security lock devices available for each rack Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-hand doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Upright Freezer Drawer racks offer a more organized system for storage and retrieval of samples. Drawer racks facilitate both access to and storage of samples without having to remove the whole rack from the freezer. Dimensions (D x H x W): 27  x 14 1/8 x 5 1/2 inches (687 x 359 x 140mm)Compartment Dimensions: 5 3/8  x 2 1/16x 5 1/4inches (137.4 x 52 x 133mm)