Learn More
Alkali Scientific Stainless Steel Freezer Racks
SDP
Supplier:  Alkali Scientific CF4L375
Encompass
The Vertical Rack for Large 3 75-inch High Boxes is a specially designed storage solution tailored for accommodating larger storage boxes within laboratory freezers including chest and upright models as well as liquid nitrogen tanks This rack is crafted to meet the specific needs of laboratories that require additional height clearance for their unique storage boxes ensuring both optimal space utilization and easy access to stored samples Product Features Specially Designed for Large Boxes This rack is specifically engineered to hold boxes up to 3 75 inches in height providing the perfect fit for larger storage containers that are commonly used for bulk or oversized samples Durable Construction Made from materials selected for their strength and durability this rack is built to withstand the extreme temperatures and conditions of laboratory freezers and liquid nitrogen tanks Its construction ensures long-term reliability and stability for your stored samples Efficient Space Utiliz
Catalog No. 50-302-6533