Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Freezer Racks for Large 3.75" Boxes, 4 x 2 Configuration, 8 Box Capacity
SDP
Catalog No. 503027848
Encompass_Preferred
Upright Freezer Racks These Upright Freezer Racks for 3.75" standard boxes offer convenient storage of samples in an organized format. Constructed of corrosion resistant stainless steel, these freezer racks include durable handles for an easy removal from freezer. Dimensions L x H x W: 24 5/16 x 8 1/16 x 5 15/16 (315.5 x 205 x 151mm)Compartment Dimensions: 5 7/8 x 4 x 5 13/16 inch (149 x 101 x 148mm)
Catalog No. 50-302-7848 Supplier Alkali Scientific Supplier No. UFR42L375
Upright Freezer Racks These Upright Freezer Racks for 3.75" standard boxes offer convenient storage of samples in an organized format. Constructed of corrosion resistant stainless steel, these freezer racks include durable handles for an easy removal from freezer. Dimensions L x H x W: 24 5/16 x 8 1/16 x 5 15/16 (315.5 x 205 x 151mm)Compartment Dimensions: 5 7/8 x 4 x 5 13/16 inch (149 x 101 x 148mm)