Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Freezer Racks for Large 3.75" Boxes, 4 x 4 Configuration, 16 Box Capacity
SDP
Catalog No. 503027969
Encompass_Preferred
Constructed of corrosion resistant stainless steel Racks fit standard 3.75" cardboard, plastic, or metal boxes Security lock devices available for each rack 4 Long x 4 Tall= Holds 16 Boxes  Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-handed doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions L x H x W: 24 5/16 x 16 1/16 x 5 15/16 (315.5 x 205 x 151mm)Compartment Dimensions: 5 7/8 x 4 x 5 13/16 inch (149 x 101 x 148mm)
Catalog No. 50-302-7969 Supplier Alkali Scientific Supplier No. UFR44L375
Constructed of corrosion resistant stainless steel Racks fit standard 3.75" cardboard, plastic, or metal boxes Security lock devices available for each rack 4 Long x 4 Tall= Holds 16 Boxes  Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-handed doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions L x H x W: 24 5/16 x 16 1/16 x 5 15/16 (315.5 x 205 x 151mm)Compartment Dimensions: 5 7/8 x 4 x 5 13/16 inch (149 x 101 x 148mm)