Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Freezer Racks for 3.75" Boxes, 4 x 2 Configuration, 8 Box Capacity
SDP
Catalog No. 503027970
Encompass_Preferred
For 3.75″ High Standard BoxesCapacity Box – 4 x 2 = 8 Standard upright freezer racks for storage of 3.75-inch boxes, features 4 boxes deep by 2 boxes high configuration for a total storage of 8 boxes per rack. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. This freezer rack does not come standard with storage boxes. Dimensions L x H x W: 22 1/8 x 8 1/16 x 5 1/2 inch (562 x 205 x 140mm)Compartment Size L x H x W: 5 3/8 x 3 15/16 x 5 3/8 inch (136 x 100 x137mm)
Catalog No. 50-302-7970 Supplier Alkali Scientific Supplier No. UFR42375
For 3.75″ High Standard BoxesCapacity Box – 4 x 2 = 8 Standard upright freezer racks for storage of 3.75-inch boxes, features 4 boxes deep by 2 boxes high configuration for a total storage of 8 boxes per rack. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. This freezer rack does not come standard with storage boxes. Dimensions L x H x W: 22 1/8 x 8 1/16 x 5 1/2 inch (562 x 205 x 140mm)Compartment Size L x H x W: 5 3/8 x 3 15/16 x 5 3/8 inch (136 x 100 x137mm)