Learn More
Alkali Scientific Stainless Steel Freezer Racks
SDP
Catalog No. 503026424
Encompass_Preferred
Freezer Storage Racks are constructed of high-quality corrosion resistant stainless steel Handles at both ends accommodate freezers with right or left-hand doors Our freezer racks have been configured to fit all major ultra-low (-80) freezers on the market racks are designed for ultra-low upright freezers Vertical Racks for Chest Freezers and Liquid Nitrogen Tanks are made of corrosion resistant stainless steel Racks are made for standard 2in boxes All racks include stainless steel locking rods Constructed of corrosion resistant stainless steel Racks fit standard 2in cardboard plastic or metal boxes Security lock devices available for each rack Made with Spring Clips Dimensions L x H x W 31 1/16 x 5 5/8 x 5 1/2 ( 789 x 143 x 140mm)Compartment Size L x H x W 2 3/16 x 5 5/16 x 5 5/16 (55 x 143 5 x 135m) CF-14-2-W-XX Racks w/ 2 Box and Divider To order replace XX with either 81 or 100 cell divider Lock device is ordered separately w10 22LB(0 1kg) for cardboard box 0 37lb(0 17kg
Catalog No. 50-302-6424 Supplier Alkali Scientific Supplier No. CF142S
Freezer Storage Racks are constructed of high-quality corrosion resistant stainless steel Handles at both ends accommodate freezers with right or left-hand doors Our freezer racks have been configured to fit all major ultra-low (-80) freezers on the market racks are designed for ultra-low upright freezers Vertical Racks for Chest Freezers and Liquid Nitrogen Tanks are made of corrosion resistant stainless steel Racks are made for standard 2in boxes All racks include stainless steel locking rods Constructed of corrosion resistant stainless steel Racks fit standard 2in cardboard plastic or metal boxes Security lock devices available for each rack Made with Spring Clips Dimensions L x H x W 31 1/16 x 5 5/8 x 5 1/2 ( 789 x 143 x 140mm)Compartment Size L x H x W 2 3/16 x 5 5/16 x 5 5/16 (55 x 143 5 x 135m) CF-14-2-W-XX Racks w/ 2 Box and Divider To order replace XX with either 81 or 100 cell divider Lock device is ordered separately w10 22LB(0 1kg) for cardboard box 0 37lb(0 17kg